Thermo Scientific Chemicals

3,5-Di-tert-butyl-4-hydroxyphenylpropionic acid, 98%, Thermo Scientific Chemicals

Catalog number: B20412.06
5 g, Each
Thermo Scientific Chemicals

3,5-Di-tert-butyl-4-hydroxyphenylpropionic acid, 98%, Thermo Scientific Chemicals

Catalog number: B20412.06
5 g, Each
Quantity
Catalog number: B20412.06
also known as B20412-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
20170-32-5
IUPAC Name
3-(3,5-di-tert-butyl-4-hydroxyphenyl)propanoate
Molecular Formula
C17H25O3
InChI Key
WPMYUUITDBHVQZ-UHFFFAOYSA-M
SMILES
CC(C)(C)C1=CC(CCC([O-])=O)=CC(=C1O)C(C)(C)C
Appearance (Color)
White to cream or yellow to pale orange
Form
Crystals or powder or crystalline powder
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Identification (FTIR)
Conforms
Melting Point (clear melt)
169.5-175.5?C

Description

It is employed as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as an active pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text