Thermo Scientific Chemicals

3,5-Dinitrosalicylaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: B20424.03
1 g, Each
Thermo Scientific Chemicals

3,5-Dinitrosalicylaldehyde, 98%, Thermo Scientific Chemicals

Catalog number: B20424.03
1 g, Each
Quantity
Catalog number: B20424.03
also known as B20424-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2460-59-5
IUPAC Name
2-formyl-4,6-dinitrobenzen-1-olate
Molecular Formula
C7H3N2O6
InChI Key
FLJXIBHYDIMYRS-UHFFFAOYSA-M
SMILES
[O-]C1=C(C=O)C=C(C=C1[N+]([O-])=O)[N+]([O-])=O
Form
Powder or crystalline powder
Appearance (Color)
Pale yellow to yellow to orange or pale brown
Assay (GC)
≥97.5%
Proton NMR
Conforms to structure

Description

It is used to produce 3-(2-hydroxy-3,5-dinitro-phenyl)-propenal with acetaldehyde. 3,5-Dinitrosalicylaldehyde has been used in the preparation of salicyldimine ligand via Schiff base condensation with allyl-substituted aniline, 3-hydroxycoumarins2, chromogenic proteinase substrates and ruthenium(II) chiral Schiff base complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to produce 3-(2-hydroxy-3,5-dinitro-phenyl)-propenal with acetaldehyde. 3,5-Dinitrosalicylaldehyde has been used in the preparation of salicyldimine ligand via Schiff base condensation with allyl-substituted aniline, 3-hydroxycoumarins2, chromogenic proteinase substrates and ruthenium(II) chiral Schiff base complexes.

Solubility
Soluble in water, 0.6 g/100 mL (18°C).

Notes
Air sensitive. Store away from oxidizing agents and air. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text