3,3,3-Trifluoropropionic acid, 98%, Thermo Scientific Chemicals
3,3,3-Trifluoropropionic acid, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3,3,3-Trifluoropropionic acid, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number B20428.14
also known as B20428-14
Price (USD)/ Each
438.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
438.00
Add to cart
3,3,3-Trifluoropropionic acid, 98%, Thermo Scientific Chemicals
Catalog numberB20428.14
Price (USD)/ Each
438.00
-
Add to cart
Chemical Identifiers
CAS2516-99-6
IUPAC Name3,3,3-trifluoropropanoate
Molecular FormulaC3H2F3O2
InChI KeyKSNKQSPJFRQSEI-UHFFFAOYSA-M
SMILES[O-]C(=O)CC(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.3320-1.3360 @ 20?C
Assay (GC)≥97.5%
Appearance (Color)Clear colorless
FormLiquid
Assay (Aqueous acid-base Titration)≥97.5 to ≤102.5%
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only