4-Acetamidobenzaldehyde, 98%, Thermo Scientific Chemicals
4-Acetamidobenzaldehyde, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Acetamidobenzaldehyde, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number B20430.22
also known as B20430-22
Price (USD)/ Each
401.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
401.00
Add to cart
4-Acetamidobenzaldehyde, 98%, Thermo Scientific Chemicals
Catalog numberB20430.22
Price (USD)/ Each
401.00
-
Add to cart
Chemical Identifiers
CAS122-85-0
IUPAC NameN-(4-formylphenyl)acetamide
Molecular FormulaC9H9NO2
InChI KeySKLUWKYNZNXSLX-UHFFFAOYSA-N
SMILESCC(=O)NC1=CC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Assay (GC)≥97.5% (UK-sourced material)
CommentMaterial sourced in UK and US
Appearance (Color)White to cream to yellow
Melting Point (clear melt)153.0-159.0?C (UK-sourced material)
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only