3-Ethylcyclopentanone, is used as a chemical reagent, pharmaceutical reagent and pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Ethylcyclopentanone, is used as a chemical reagent, pharmaceutical reagent and pharmaceutical intermediate.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only
L Su.; X Li.; WL Chan.; X Jia.; ASC Chan. Copper-catalyzed enantioselective conjugate addition of triethylaluminum to 2-cyclopentenone. Tetrahedron: Asymmetry. 200314 (13), 1865-1869.
EJ Corey.; R Naef.; FJ Hannon. Enantioselective conjugate addition of rationally designed chiral cuprate reagents to 2-cycloalkenones. J. Am. Chem. Soc. 1986108 (22), 7114-7116.