Thermo Scientific Chemicals

2-(2-Naphthyl)indole, 98%, Thermo Scientific Chemicals

Catalog number: B20434.06
5 g, Each
Thermo Scientific Chemicals

2-(2-Naphthyl)indole, 98%, Thermo Scientific Chemicals

Catalog number: B20434.06
5 g, Each
Quantity
Catalog number: B20434.06
also known as B20434-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
23746-81-8
Melting Point (clear melt)
199.5-205.5?C
Appearance (Color)
Pale brown
Assay (GC)
≥97.5%
Form
Powder

Description

It is used in the catalytic enantioselective addition of indoles to arylnitroalkenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the catalytic enantioselective addition of indoles to arylnitroalkenes.

Solubility
Soluble in DMSO.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text