(3-Chloropropyl)benzene, 98%, Thermo Scientific Chemicals
(3-Chloropropyl)benzene, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(3-Chloropropyl)benzene, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number B20445.06
also known as B20445-06
Price (USD)/ Each
27.60
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
27.60
Add to cart
(3-Chloropropyl)benzene, 98%, Thermo Scientific Chemicals
Catalog numberB20445.06
Price (USD)/ Each
27.60
-
Add to cart
Chemical Identifiers
CAS104-52-9
IUPAC Name(3-chloropropyl)benzene
Molecular FormulaC9H11Cl
InChI KeyXZBXAYCCBFTQHH-UHFFFAOYSA-N
SMILESClCCCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Refractive Index1.5205-1.5245 @ 20°C
Appearance (Color)Clear colorless to yellow to brown
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only