Thermo Scientific Chemicals

2-(4-Pyridyl)thiazole-4-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: B20457.03
1 g, Each
Thermo Scientific Chemicals

2-(4-Pyridyl)thiazole-4-carboxylic acid, 97%, Thermo Scientific Chemicals

Catalog number: B20457.03
1 g, Each
Quantity
Catalog number: B20457.03
also known as B20457-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
21278-86-4
IUPAC Name
2-(pyridin-4-yl)-1,3-thiazole-4-carboxylate
Molecular Formula
C9H5N2O2S
InChI Key
COOQMBOJAAZEIR-UHFFFAOYSA-M
SMILES
[O-]C(=O)C1=CSC(=N1)C1=CC=NC=C1
Form
Powder
Assay (Aqueous acid-base Titration)
≥96.0 to ≤104.0%
Appearance (Color)
Cream to pale brown
Assay (Silylated GC)
≥96.0%

Description

2-(4-Pyridyl)thiazole-4-carboxylic acid, is used as pharmaceutical intermediate and also used in the field of organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(4-Pyridyl)thiazole-4-carboxylic acid, is used as pharmaceutical intermediate and also used in the field of organic synthesis.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under normal temperatures and pressures. Keep away from strong oxidizing agents, strong acids, strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text