Thermo Scientific Chemicals

N.N'-Bis(2-hydroxyethyl)oxamide, 99%, Thermo Scientific Chemicals

Catalog number: B20458.06
5 g, Each
Thermo Scientific Chemicals

N.N'-Bis(2-hydroxyethyl)oxamide, 99%, Thermo Scientific Chemicals

Catalog number: B20458.06
5 g, Each
Quantity
Catalog number: B20458.06
also known as B20458-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1871-89-2
IUPAC Name
N,N'-bis(2-hydroxyethyl)ethanediamide
Molecular Formula
C6H12N2O4
InChI Key
FPQJEXTVQZHURJ-UHFFFAOYSA-N
SMILES
OCCNC(=O)C(=O)NCCO
Appearance (Color)
White
Form
Crystalline powder
Assay (GC)
>98.5%

Description

N.N'-Bis(2-hydroxyethyl)oxamide is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N.N′-Bis(2-hydroxyethyl)oxamide is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under normal temperatures and pressures. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text