Thermo Scientific Chemicals

Ethyl bromodifluoroacetate, 97%, Thermo Scientific Chemicals

Catalog number: B20461.06
5 g, Each
Thermo Scientific Chemicals

Ethyl bromodifluoroacetate, 97%, Thermo Scientific Chemicals

Catalog number: B20461.06
5 g, Each
Quantity
Catalog number: B20461.06
also known as B20461-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
667-27-6
IUPAC Name
ethyl 2-bromo-2,2-difluoroacetate
Molecular Formula
C4H5BrF2O2
InChI Key
IRSJDVYTJUCXRV-UHFFFAOYSA-N
SMILES
CCOC(=O)C(F)(F)Br
Assay (GC)
≥96.0%
Refractive Index
1.3840-1.3890 @ 20?C
Appearance (Color)
Clear colorless
Form
Liquid

Description

Ethyl bromodifluoroacetate, is used for the treatment of the Reformat sky reagent with aldehydes and ketones affords 2,2-difluoro-3-hydroxy esters. It is also used in the analysis of reagent purity through IR, NMR.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl bromodifluoroacetate, is used for the treatment of the Reformat sky reagent with aldehydes and ketones affords 2,2-difluoro-3-hydroxy esters. It is also used in the analysis of reagent purity through IR, NMR.

Solubility
Soluble in most organic solvents.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text