Thermo Scientific Chemicals

1-Bromo-5-chloropentane, 98%, Thermo Scientific Chemicals

Catalog number: B20479.09
10 g, Each
Thermo Scientific Chemicals

1-Bromo-5-chloropentane, 98%, Thermo Scientific Chemicals

Catalog number: B20479.09
10 g, Each
Quantity
Catalog number: B20479.09
also known as B20479-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
54512-75-3
IUPAC Name
1-bromo-5-chloropentane
Molecular Formula
C5H10BrCl
InChI Key
PHHNNDKXQVKJEP-UHFFFAOYSA-N
SMILES
ClCCCCCBr
Form
Liquid
Appearance (Color)
Clear colorless
Assay (GC)
≥97.5%
Refractive Index
1.4825-1.4865 @ 20?C

Description

1-Bromo-5-chloropentane, is used as an primary and secondary intermediate. It is also used as a intermediate in organic syntheses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-5-chloropentane, is used as an primary and secondary intermediate. It is also used as a intermediate in organic syntheses.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under normal temperatures and pressures. Keep away from strong oxidizing agents, strong acids, strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text