2-Fluoro-4-(trifluoromethyl)benzonitrile, 98%, Thermo Scientific Chemicals
2-Fluoro-4-(trifluoromethyl)benzonitrile, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-Fluoro-4-(trifluoromethyl)benzonitrile, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B20485.03
also known as B20485-03
Price (USD)/ Each
55.10
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
55.10
Add to cart
2-Fluoro-4-(trifluoromethyl)benzonitrile, 98%, Thermo Scientific Chemicals
Catalog numberB20485.03
Price (USD)/ Each
55.10
-
Add to cart
Chemical Identifiers
CAS146070-34-0
IUPAC Name2-fluoro-4-(trifluoromethyl)benzonitrile
Molecular FormulaC8H3F4N
InChI KeyJLTYVTXTSOYXMX-UHFFFAOYSA-N
SMILESFC1=CC(=CC=C1C#N)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay (GC)≥97.5%
FormLiquid
Refractive Index1.4450-1.4490 @ 20?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only