Thermo Scientific Chemicals

2,6-Bis(trifluoromethyl)benzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: B20486.06
5 g, Each
Thermo Scientific Chemicals

2,6-Bis(trifluoromethyl)benzoic acid, 98%, Thermo Scientific Chemicals

Catalog number: B20486.06
5 g, Each
Quantity
Catalog number: B20486.06
also known as B20486-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
24821-22-5
IUPAC Name
2,6-bis(trifluoromethyl)benzoic acid
Molecular Formula
C9H4F6O2
InChI Key
XZNLSDPNMNWCRE-UHFFFAOYSA-N
SMILES
OC(=O)C1=C(C=CC=C1C(F)(F)F)C(F)(F)F
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
134.0-140.0?C
Appearance (Color)
White to pale cream to pale yellow
Assay (Silylated GC)
≥97.5%
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%

Description

2,6-Bis(trifluoromethyl)benzoic acid is used as an organic intermediate in synthesis. It is also used as a pharmaceutical intermediate and organic intermediate in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Bis(trifluoromethyl)benzoic acid is used as an organic intermediate in synthesis. It is also used as a pharmaceutical intermediate and organic intermediate in chemical synthesis.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text