Ursodeoxycholic acid, 99%, Thermo Scientific Chemicals
Ursodeoxycholic acid, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ursodeoxycholic acid, 99%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B20490.03
also known as B20490-03
Price (USD)/ Each
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS128-13-2
IUPAC Name(4R)-4-[(1R,3aS,3bR,4S,5aS,7R,9aS,9bS,11aR)-4,7-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoic acid
Molecular FormulaC24H40O4
InChI KeyRUDATBOHQWOJDD-UZVSRGJWSA-N
SMILES[H][C@@]12CC[C@H]([C@H](C)CCC(O)=O)[C@@]1(C)CC[C@@]1([H])[C@@]2([H])[C@@H](O)C[C@]2([H])C[C@H](O)CC[C@]12C
View more
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)200.0-206.0?C
FormCrystals or powder or crystalline powder
Optical Rotation+60.0 ± 2.0? (C=2 in ethanol)
Appearance (Color)White
Assay (Aqueous acid-base Titration)≥98.5 to ≤101.5%
A secondary bile acid

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only