4-Chlorostyrene, 98+%, stab, Thermo Scientific Chemicals
4-Chlorostyrene, 98+%, stab, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Chlorostyrene, 98+%, stab, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
Catalog number B20512.09
also known as B20512-09
Price (USD)/ Each
77.10
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
77.10
Add to cart
4-Chlorostyrene, 98+%, stab, Thermo Scientific Chemicals
Catalog numberB20512.09
Price (USD)/ Each
77.10
-
Add to cart
Chemical Identifiers
CAS1073-67-2
IUPAC Name1-chloro-4-ethenylbenzene
Molecular FormulaC8H7Cl
InChI KeyKTZVZZJJVJQZHV-UHFFFAOYSA-N
SMILESClC1=CC=C(C=C)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Identification (FTIR)Conforms
Refractive Index1.5640-1.5670 @ 20°C
Solution Test10% in Diethyl ether: Clear
Solution Test10% in Methanol: Clear
Appearance (Color)Clear colorless to yellow to orange
View more
4-Chlorostyrene is used in the preparation of new bis(pyrazolyl)borato olefin complexes of copper(I). It is also employed in the investigation of regioselectivity in the cationic Heck reaction of 4-substituted styrenes. Further, it plays an important role in the preparation of 4-[(4-chloro-phenyl)-thioacetyl]-morpholine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorostyrene is used in the preparation of new bis(pyrazolyl)borato olefin complexes of copper(I). It is also employed in the investigation of regioselectivity in the cationic Heck reaction of 4-substituted styrenes. Further, it plays an important role in the preparation of 4-[(4-chloro-phenyl)-thioacetyl]-morpholine.

Solubility
Immiscible with water.

Notes
Incompatible with acids, bases, oxidizing agents, halogens, metal salts and polymerizing initiators. Store in a cool place.
RUO – Research Use Only

General References:

  1. Dhakshinamoorthy, A.; Latorre-Sanchez, M.; Asiri, A. M.; Primo, A.; Garcia, H. Sulphur-doped graphene as metal-free carbocatalysts for the solventless aerobic oxidation of styrenes. Catal. Commun. 2015, 65, 10-13.
  2. Wang, B.; Lee, Y. M.; Seo, M. S.; Nam, W. Mononuclear Nonheme Iron(III)-Iodosylarene and High-Valent Iron-Oxo Complexes in Olefin Epoxidation Reactions. Angew. Chem. Int. Ed. 2015, 54 (40), 11740-11744.