Thermo Scientific Chemicals

4-Chlorostyrene, 98+%, stab, Thermo Scientific Chemicals

Catalog number: B20512.18
50 g, Each
Thermo Scientific Chemicals

4-Chlorostyrene, 98+%, stab, Thermo Scientific Chemicals

Catalog number: B20512.18
50 g, Each
Quantity
Catalog number: B20512.18
also known as B20512-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1073-67-2
IUPAC Name
1-chloro-4-ethenylbenzene
Molecular Formula
C8H7Cl
InChI Key
KTZVZZJJVJQZHV-UHFFFAOYSA-N
SMILES
ClC1=CC=C(C=C)C=C1
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to yellow to orange
Form
Liquid
Assay (GC)
≥98.0%
Refractive Index
1.5640-1.5670 @ 20°C

Description

4-Chlorostyrene is used in the preparation of new bis(pyrazolyl)borato olefin complexes of copper(I). It is also employed in the investigation of regioselectivity in the cationic Heck reaction of 4-substituted styrenes. Further, it plays an important role in the preparation of 4-[(4-chloro-phenyl)-thioacetyl]-morpholine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Chlorostyrene is used in the preparation of new bis(pyrazolyl)borato olefin complexes of copper(I). It is also employed in the investigation of regioselectivity in the cationic Heck reaction of 4-substituted styrenes. Further, it plays an important role in the preparation of 4-[(4-chloro-phenyl)-thioacetyl]-morpholine.

Solubility
Immiscible with water.

Notes
Incompatible with acids, bases, oxidizing agents, halogens, metal salts and polymerizing initiators. Store in a cool place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text