1,5-Dibromo-3-methylpentane is used as a chemical reagent, pharmaceutical intermediate, pharmaceutical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,5-Dibromo-3-methylpentane is used as a chemical reagent, pharmaceutical intermediate, pharmaceutical research.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, strong bases.
RUO – Research Use Only
General References:
- NJ Leonard.; ZW Wicks. 1, 5-Dibromo-3-methylpentane. J. Am. Chem. Soc. 194668 (11), 2402-2404.
- CR Johnson.; D McCants. The Stereochemistry of Oxidation at Sulfur. Oxidation of 4-Substituted Thianes to Sulfoxides1. J. Am. Chem. Soc. 196587 (5), 1109-1114.