Thermo Scientific Chemicals

Methyl 5-bromo-2-furoate, 97%, Thermo Scientific Chemicals

Catalog number: B20524.03
1 g, Each
Thermo Scientific Chemicals

Methyl 5-bromo-2-furoate, 97%, Thermo Scientific Chemicals

Catalog number: B20524.03
1 g, Each
Quantity
Catalog number: B20524.03
also known as B20524-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2527-99-3
IUPAC Name
methyl 5-bromofuran-2-carboxylate
Molecular Formula
C6H5BrO3
InChI Key
FBPIDMAELBIRLE-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC=C(Br)O1
Appearance (Color)
White
Assay (GC)
≥96.0%
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
62.0-72.0?C

Description

It is used in the preparation of hydroquinone bis(2-carbomethoxy-5-furyl ether).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the preparation of hydroquinone bis(2-carbomethoxy-5-furyl ether).

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text