Methyl 5-bromo-2-furoate, 97%, Thermo Scientific Chemicals
Methyl 5-bromo-2-furoate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 5-bromo-2-furoate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number B20524.06
also known as B20524-06
Price (USD)/ Each
239.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
239.00
Add to cart
Methyl 5-bromo-2-furoate, 97%, Thermo Scientific Chemicals
Catalog numberB20524.06
Price (USD)/ Each
239.00
-
Add to cart
Chemical Identifiers
CAS2527-99-3
IUPAC Namemethyl 5-bromofuran-2-carboxylate
Molecular FormulaC6H5BrO3
InChI KeyFBPIDMAELBIRLE-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(Br)O1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Melting Point (clear melt)62.0-72.0?C
Appearance (Color)White
Assay (GC)≥96.0%
It is used in the preparation of hydroquinone bis(2-carbomethoxy-5-furyl ether).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the preparation of hydroquinone bis(2-carbomethoxy-5-furyl ether).

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

General References:

  1. Donald G. Manly, et al. Simple Furan Ethers. II: 2-Alkoxy- and 2-Aryloxy-furans.J. Org. Chem.,1956,21(5), 516-519.
  2. James A. Cella. 5-Substituted-2-furoic acids as latent dienes for the preparation of aryl ethers and thioethers via the Diels-Alder reaction.J. Org. Chem.,1988,59(3), 2099-2103.