Thermo Scientific Chemicals

3',5,7-Trihydroxy-4'-methoxyflavanone, 97%, Thermo Scientific Chemicals

Catalog number: B20528.14
25 g, Each
Thermo Scientific Chemicals

3',5,7-Trihydroxy-4'-methoxyflavanone, 97%, Thermo Scientific Chemicals

Catalog number: B20528.14
25 g, Each
Quantity
Catalog number: B20528.14
also known as B20528-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
520-33-2
IUPAC Name
(2S)-5,7-dihydroxy-2-(3-hydroxy-4-methoxyphenyl)-3,4-dihydro-2H-1-benzopyran-4-one
Molecular Formula
C16H14O6
InChI Key
AIONOLUJZLIMTK-AWEZNQCLSA-N
SMILES
COC1=CC=C(C=C1O)[C@@H]1CC(=O)C2=C(O)C=C(O)C=C2O1
Appearance (Color)
White to cream or pale yellow to pale brown
Form
Powder
Identification (FTIR)
Conforms
Assay (HPLC)
≥96.0%
Melting Point (clear melt)
225-236?C

Description

(±)-Hesperetin is an antioxidant flavonoid. Induces G1-phase cell cycle arrest. Anti-inflammatory. (±)-Hesperetin suppresses NF-κB activation. Reduces cholesterol biosynthesis. Inhibits lipid peroxidation. Neuroprotective against neuronal oxidative damage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(±)-Hesperetin is an antioxidant flavonoid. Induces G1-phase cell cycle arrest. Anti-inflammatory. (±)-Hesperetin suppresses NF-κB activation. Reduces cholesterol biosynthesis. Inhibits lipid peroxidation. Neuroprotective against neuronal oxidative damage.

Solubility
Soluble in water (partly), dilute alkalis, and ethanol (50 mg/ml).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text