Thermo Scientific Chemicals

2-Mercaptopyridine N-oxide sodium salt, anhydrous, 98%, Thermo Scientific Chemicals

Catalog number: B20529.03
1 g, Each
Thermo Scientific Chemicals

2-Mercaptopyridine N-oxide sodium salt, anhydrous, 98%, Thermo Scientific Chemicals

Catalog number: B20529.03
1 g, Each
Quantity
Catalog number: B20529.03
also known as B20529-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
3811-73-2
IUPAC Name
sodium 2-sulfanylidene-1,2-dihydropyridin-1-olate
Molecular Formula
C5H4NNaOS
InChI Key
XNRNJIIJLOFJEK-UHFFFAOYSA-N
SMILES
[Na+].[O-]N1C=CC=CC1=S
Appearance (Color)
Pale cream to cream
Form
Crystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%

Description

2-Mercaptopyridine N-oxide sodium salt is one of the active components in paint, sealants, shampoo, adhesive and aerosol due to its anti-microbial activity. In biochemistry studies, it is utilized to transport zinc into cells. Further, it is used to form bidentate oxothiolane chelates with transition metals. It acts as a stabilizer and viscosity building provider in weak basic or neutral medium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Mercaptopyridine N-oxide sodium salt is one of the active components in paint, sealants, shampoo, adhesive and aerosol due to its anti-microbial activity. In biochemistry studies, it is utilized to transport zinc into cells. Further, it is used to form bidentate oxothiolane chelates with transition metals. It acts as a stabilizer and viscosity building provider in weak basic or neutral medium.

Solubility
Soluble in water, ethanol, propylene glycol, polyethylene glycol and dimethyl sulfoxide. Insoluble in liquid paraffin and olive oil.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text