Thermo Scientific Chemicals

N-Methylanthranilic acid, 90+%, Thermo Scientific Chemicals

Catalog number: B20531.14
25 g, Each
Thermo Scientific Chemicals

N-Methylanthranilic acid, 90+%, Thermo Scientific Chemicals

Catalog number: B20531.14
25 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: B20531.14
also known as B20531-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
119-68-6
IUPAC Name
2-(methylamino)benzoic acid
Molecular Formula
C8H9NO2
InChI Key
WVMBPWMAQDVZCM-UHFFFAOYSA-N
SMILES
CNC1=CC=CC=C1C(O)=O
Assay (Non-aqueous acid-base Titration)
>90.0% (total base)
Assay (Aqueous acid-base Titration)
>90.0%
Form
powder
Appearance (Color)
Brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text