Thermo Scientific Chemicals

2-Fluoro-3-methylbenzyl bromide, 97%, Thermo Scientific Chemicals

Catalog number: B20538.03
1 g, Each
Thermo Scientific Chemicals

2-Fluoro-3-methylbenzyl bromide, 97%, Thermo Scientific Chemicals

Catalog number: B20538.03
1 g, Each
Quantity
Catalog number: B20538.03
also known as B20538-03
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
151412-12-3
IUPAC Name
1-(bromomethyl)-2-fluoro-3-methylbenzene
Molecular Formula
C8H8BrF
InChI Key
DCGWFNZSJBIAKE-UHFFFAOYSA-N
SMILES
CC1=C(F)C(CBr)=CC=C1
Appearance (Color)
Clear colorless
Refractive Index
1.5450-1.5500 @ 20?C
Form
Liquid
Assay (GC)
≥96.0%

Description

2-Fluoro-3-methylbenzyl bromide, is used as a chemical and organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoro-3-methylbenzyl bromide, is used as a chemical and organic intermediate.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, strong bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text