2-Fluoro-3-methylbenzyl bromide, is used as a chemical and organic intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Fluoro-3-methylbenzyl bromide, is used as a chemical and organic intermediate.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, strong bases.
RUO – Research Use Only
General References:
- J Jiang.; MB Boxer.; MG Vander Heiden.; M Shen. Evaluation of thieno [3, 2-b] pyrrole [3, 2-d] pyridazinones as activators of the tumor cell specific M2 isoform of pyruvate kinase. Bioorganic & Medicinal Chemistry Letters. 201020 (11), 3387-3393.
- H Plenio.; R Diodone. Covalently bonded fluorine as a σ-donor for groups I and II metal ions in partially fluorinated macrocycles. J. Am. Chem. Soc. 1996118 (2), 356-367.