Thermo Scientific Chemicals

o-Toluenesulfonamide, 98%, Thermo Scientific Chemicals

Catalog number: B20544.18
50 g, Each
Thermo Scientific Chemicals

o-Toluenesulfonamide, 98%, Thermo Scientific Chemicals

Catalog number: B20544.18
50 g, Each
Quantity
Catalog number: B20544.18
also known as B20544-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
88-19-7
IUPAC Name
2-methylbenzene-1-sulfonamide
Molecular Formula
C7H9NO2S
InChI Key
YCMLQMDWSXFTIF-UHFFFAOYSA-N
SMILES
CC1=CC=CC=C1S(N)(=O)=O
Appearance (Color)
White to cream
Assay (GC)
≥97.5%
Form
Crystals or powder or crystalline powder
Melting Point (clear melt)
153.0-160.0°C

Description

o-Toluene sulfonamide was mainly produced as a chemical intermediate for the production of saccharin. A mixture of o-Toluene sulfonamide with the p-isomer is used as a plasticizer for hot -melt adhesives, a chemical intermediate for fluorescent pigments and a chemical intermediate for plasticizer resins. (S080800). Studies suggest that o-Toluene sulfonamide has potential mutagenic activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
o-Toluene sulfonamide was mainly produced as a chemical intermediate for the production of saccharin. A mixture of o-Toluene sulfonamide with the p-isomer is used as a plasticizer for hot -melt adhesives, a chemical intermediate for fluorescent pigments and a chemical intermediate for plasticizer resins. (S080800). Studies suggest that o-Toluene sulfonamide has potential mutagenic activity.

Solubility
Soluble in water (1.6 g/L at 25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, bases and acids.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text