Methyl 3-methoxythiophene-2-carboxylate, 97%, Thermo Scientific Chemicals
Methyl 3-methoxythiophene-2-carboxylate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Methyl 3-methoxythiophene-2-carboxylate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number B20586.06
also known as B20586-06
Price (USD)/ Each
215.00
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
215.00
Add to cart
Methyl 3-methoxythiophene-2-carboxylate, 97%, Thermo Scientific Chemicals
Catalog numberB20586.06
Price (USD)/ Each
215.00
-
Add to cart
Chemical Identifiers
CAS62353-75-7
IUPAC Namemethyl 3-methoxythiophene-2-carboxylate
Molecular FormulaC7H8O3S
InChI KeyMWADWXJPBMFRGL-UHFFFAOYSA-N
SMILESCOC(=O)C1=C(OC)C=CS1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale brown
Assay (GC)≥96.0%
FormCrystals or powder or crystalline powder
Melting Point (clear melt)50.0-59.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only