2,2,3,4,4,4-Hexafluoro-1-butanol, 95%, Thermo Scientific Chemicals
2,2,3,4,4,4-Hexafluoro-1-butanol, 95%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,2,3,4,4,4-Hexafluoro-1-butanol, 95%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number B20587.14
also known as B20587-14
Price (USD)/ Each
128.00
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
128.00
Add to cart
2,2,3,4,4,4-Hexafluoro-1-butanol, 95%, Thermo Scientific Chemicals
Catalog numberB20587.14
Price (USD)/ Each
128.00
-
Add to cart
Chemical Identifiers
CAS382-31-0
IUPAC Name2,2,3,4,4,4-hexafluorobutan-1-ol
Molecular FormulaC4H4F6O
InChI KeyLVFXLZRISXUAIL-UHFFFAOYNA-N
SMILESOCC(F)(F)C(F)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Assay (GC)≥94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only