Sodium hexafluoroantimonate(V), 98%, Thermo Scientific Chemicals
Sodium hexafluoroantimonate(V), 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium hexafluoroantimonate(V), 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Catalog number B20604.14
also known as B20604-14
Price (USD)/ Each
49.80
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
49.80
Add to cart
Sodium hexafluoroantimonate(V), 98%, Thermo Scientific Chemicals
Catalog numberB20604.14
Price (USD)/ Each
49.80
-
Add to cart
Chemical Identifiers
CAS16925-25-0
Molecular FormulaF6NaSb
InChI KeyZJLVIRKMUXZEDU-UHFFFAOYSA-N
SMILES[F].[F].[F].[F].[F].[F].[Na+].[Sb]
Molecular Weight (g/mol)258.74
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay from Supplier's CofA≥97.5%
FormPowder or crystals or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only