Thermo Scientific Chemicals

Sodium hexafluoroantimonate(V), 98%, Thermo Scientific Chemicals

Catalog number: B20604.36
500 g, Each
Thermo Scientific Chemicals

Sodium hexafluoroantimonate(V), 98%, Thermo Scientific Chemicals

Catalog number: B20604.36
500 g, Each
Quantity
Catalog number: B20604.36
also known as B20604-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16925-25-0
Molecular Formula
F6NaSb
InChI Key
ZJLVIRKMUXZEDU-UHFFFAOYSA-N
SMILES
[F].[F].[F].[F].[F].[F].[Na+].[Sb]
Molecular Weight (g/mol)
258.74
Appearance (Color)
White
Assay from Supplier's CofA
≥97.5%
Form
Powder or crystals or crystalline powder

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text