Thermo Scientific Chemicals

4-Fluoro-2-(trifluoromethyl)benzonitrile, 98+%, Thermo Scientific Chemicals

Catalog number: B20617.14
25 g, Each
Thermo Scientific Chemicals

4-Fluoro-2-(trifluoromethyl)benzonitrile, 98+%, Thermo Scientific Chemicals

Catalog number: B20617.14
25 g, Each
Quantity
Catalog number: B20617.14
also known as B20617-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
194853-86-6
IUPAC Name
4-fluoro-2-(trifluoromethyl)benzonitrile
Molecular Formula
C8H3F4N
InChI Key
LCCPQUYXMFXCAC-UHFFFAOYSA-N
SMILES
FC1=CC=C(C#N)C(=C1)C(F)(F)F
Melting Point (clear melt)
43.0-49.0?C
Form
Lumps or fused solid
Appearance (Color)
White
Assay (GC)
≥98.0%

Description

It is used as a pharmaceutical intermediate. It is also used for the synthesis of diphenylthioethers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a pharmaceutical intermediate. It is also used for the synthesis of diphenylthioethers.

Solubility
Insoluble in water. Solubility in methanol is almost transparent.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text