Thermo Scientific Chemicals

2-Iodobutane, 99%, stab. with copper, Thermo Scientific Chemicals

Catalog number: B20619.18
50 g, Each
Thermo Scientific Chemicals

2-Iodobutane, 99%, stab. with copper, Thermo Scientific Chemicals

Catalog number: B20619.18
50 g, Each
Quantity
Catalog number: B20619.18
also known as B20619-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
513-48-4
Appearance (Color)
Clear colorless to orange to pale brown
Assay (GC)
≥98.5%
Refractive Index
1.4980-1.5010 @ 20?C
Form
Liquid
Identification (FTIR)
Conforms

Description

It is used as a solvent organic synthesis and as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a solvent organic synthesis and as pharmaceutical intermediate.

Solubility
Insoluble in water. Soluble in alcohol and diethyl ether.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text