Thermo Scientific Chemicals

3,4-Dimethoxyphenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: B20625.14
25 g, Each
Thermo Scientific Chemicals

3,4-Dimethoxyphenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog number: B20625.14
25 g, Each
Quantity
Catalog number: B20625.14
also known as B20625-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
93-40-3
IUPAC Name
2-(3,4-dimethoxyphenyl)acetic acid
Molecular Formula
C10H12O4
InChI Key
WUAXWQRULBZETB-UHFFFAOYSA-N
SMILES
COC1=CC=C(CC(O)=O)C=C1OC
Assay (Aqueous acid-base Titration)
≥97.5 to ≤102.5%
Form
Powder
Melting Point (clear melt)
94.0-101.0?C
Appearance (Color)
Pale cream
Assay (HPLC)
≥97.5%

Description

It reacts with formaldehyde in the presence of acid to give an isochromanone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It reacts with formaldehyde in the presence of acid to give an isochromanone.

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text