Thermo Scientific Chemicals

Cyclopentene oxide, 97%, Thermo Scientific Chemicals

Catalog number: B20629.14
25 g, Each
Thermo Scientific Chemicals

Cyclopentene oxide, 97%, Thermo Scientific Chemicals

Catalog number: B20629.14
25 g, Each
Quantity
Catalog number: B20629.14
also known as B20629-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
285-67-6
Form
Liquid
Identification (FTIR)
Conforms
Appearance (Color)
Clear colorless to pale yellow
Refractive Index
1.4320-1.4370 @ 20?C
Assay (GC)
≥96.0%

Description

Cyclopentene oxide is used to prepare beta-amino alcohols by reacting with aromatic amines using bismuth trichloride as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopentene oxide is used to prepare beta-amino alcohols by reacting with aromatic amines using bismuth trichloride as a catalyst.

Solubility
Immiscible with water.

Notes
Store in a cool place. Incompatible with acids, bases and oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text