2,5-Dimethylpyrrole-3-carboxylic acid, 97%, Thermo Scientific Chemicals
2,5-Dimethylpyrrole-3-carboxylic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2,5-Dimethylpyrrole-3-carboxylic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Catalog number B20635.03
also known as B20635-03
Price (USD)/ Each
107.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
107.00
Add to cart
2,5-Dimethylpyrrole-3-carboxylic acid, 97%, Thermo Scientific Chemicals
Catalog numberB20635.03
Price (USD)/ Each
107.00
-
Add to cart
Chemical Identifiers
CAS57338-76-8
IUPAC Name2,5-dimethyl-1H-pyrrole-3-carboxylate
Molecular FormulaC7H8NO2
InChI KeyVDVWTJFVFQVCFN-UHFFFAOYSA-M
SMILESCC1=CC(C([O-])=O)=C(C)N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream to pale brown
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only