Thermo Scientific Chemicals

5-Iodoindole, 98%, Thermo Scientific Chemicals

Catalog number: B20638.14
25 g, Each
Thermo Scientific Chemicals

5-Iodoindole, 98%, Thermo Scientific Chemicals

Catalog number: B20638.14
25 g, Each
Quantity
Catalog number: B20638.14
also known as B20638-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
16066-91-4
IUPAC Name
5-iodo-1H-indole
Molecular Formula
C8H6IN
InChI Key
TVQLYTUWUQMGMP-UHFFFAOYSA-N
SMILES
IC1=CC=C2NC=CC2=C1
Identification (FTIR)
Conforms
Appearance (Color)
White to pink or pale cream to cream to pale brown
Assay (GC)
≥96.0%
Melting Point (clear melt)
94.0-102.0°C
Form
Crystals or powder or crystalline powder

Description

5-Iodoindole is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. It is used as the starting material in the synthesis of 5-(α-fluorovinyl)-N-tosylindole.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Iodoindole is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. It is used as the starting material in the synthesis of 5-(α-fluorovinyl)-N-tosylindole.

Notes
Store at room temperature. Incompatible with strong oxidizing agents. Light sensitive.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text