Thermo Scientific Chemicals

4-(Bromomethyl)benzonitrile, 98%, Thermo Scientific Chemicals

Catalog number: B20641.06
5 g, Each
Thermo Scientific Chemicals

4-(Bromomethyl)benzonitrile, 98%, Thermo Scientific Chemicals

Catalog number: B20641.06
5 g, Each
Quantity
Catalog number: B20641.06
also known as B20641-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
17201-43-3
IUPAC Name
4-(bromomethyl)benzonitrile
Molecular Formula
C8H6BrN
InChI Key
UMLFTCYAQPPZER-UHFFFAOYSA-N
SMILES
BrCC1=CC=C(C=C1)C#N
Appearance (Color)
White to cream
Assay (GC)
≥97.5%
Form
Crystals or powder or crystalline powder or lumps
Melting Point (clear melt)
112.5-118.5?C

Description

4-(Bromomethyl)benzonitrile is used in the synthesis of ligands containing a chelating pyrazolyl-pyridine group with a pendant aromatic nitrile. It is also useful in the preparation of 4-[(2H-tetra-zol-2-yl)methyl]benzonitrile by reaction with 2H-tetrazole in the presence of potassium hydroxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-(Bromomethyl)benzonitrile is used in the synthesis of ligands containing a chelating pyrazolyl-pyridine group with a pendant aromatic nitrile. It is also useful in the preparation of 4-[(2H-tetra-zol-2-yl)methyl]benzonitrile by reaction with 2H-tetrazole in the presence of potassium hydroxide.

Solubility
Soluble in chloroform and methanol. Insoluble in water.

Notes
Moisture and light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents, strong acids and bases.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text