Ethyl 3,5-dimethylisoxazole-4-carboxylate, 97%, Thermo Scientific Chemicals
Ethyl 3,5-dimethylisoxazole-4-carboxylate, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ethyl 3,5-dimethylisoxazole-4-carboxylate, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 g
10 g
Catalog number B20642.88
also known as B20642-88
Price (USD)/ Each
47.50
-
Add to cart
Quantity:
2.5 g
Request bulk or custom format
Price (USD)/ Each
47.50
Add to cart
Ethyl 3,5-dimethylisoxazole-4-carboxylate, 97%, Thermo Scientific Chemicals
Catalog numberB20642.88
Price (USD)/ Each
47.50
-
Add to cart
Chemical Identifiers
CAS17147-42-1
IUPAC Nameethyl 3,5-dimethyl-1,2-oxazole-4-carboxylate
Molecular FormulaC8H11NO3
InChI KeyOQRHVDXUJAQVNT-UHFFFAOYSA-N
SMILESCCOC(=O)C1=C(C)ON=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
FormLiquid
Refractive Index1.4595-1.4645 @ 20?C
Appearance (Color)Clear colorless to pale yellow
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only