It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.
Solubility
Soluble in water.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
- Yasuto Tsuruta.; Kazuya Kohashi. Sensitive derivatization reagents for hydroxyl and amino compounds for thin-layer or high-performance liquid chromatography with fluorescence detection. Anal. Chim. Acta. 1987, 192 309-313 .
- Manuel J. S. Monte.; Ana R. R. P. Almeida.; M. Agostinha R. Matos. Thermodynamic Study on the Sublimation of Five Aminomethoxybenzoic Acids.J. Chem. Eng. Data. 2010, 55 (1),419-423 .