Thermo Scientific Chemicals

Phenyl salicylate, 99%, Thermo Scientific Chemicals

Catalog number: B20686.22
100 g, Each
Thermo Scientific Chemicals

Phenyl salicylate, 99%, Thermo Scientific Chemicals

Catalog number: B20686.22
100 g, Each
Quantity
Catalog number: B20686.22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
118-55-8
IUPAC Name
phenyl 2-hydroxybenzoate
Molecular Formula
C13H10O3
InChI Key
ZQBAKBUEJOMQEX-UHFFFAOYSA-N
SMILES
OC1=CC=CC=C1C(=O)OC1=CC=CC=C1
Assay (Silylated GC)
≥98.5%
Form
Crystals or powder or crystalline powder or fused solid
Melting Point (clear melt)
38.0-45.0?C
Appearance (Color)
White
Identification (FTIR)
Conforms

Description

Phenyl salicylate is used in the production of polymers, lacquers, adhesives, waxes and polishes. It acts as an antiseptic and mild analgesic. It is also used in the prevention of sunburn. It reacts with o-toluidine in 1,2,4-trichlorobenzene to give the corresponding amide o-Salicylotoluide. Further, it is involved in the study to check the effect on crystal size with cooling rate in the laboratory. In addition to this, it is used in veterinary medicine and also employed for coating pills, in which the medicine is intended for enteric release salol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phenyl salicylate is used in the production of polymers, lacquers, adhesives, waxes and polishes. It acts as an antiseptic and mild analgesic. It is also used in the prevention of sunburn. It reacts with o-toluidine in 1,2,4-trichlorobenzene to give the corresponding amide o-Salicylotoluide. Further, it is involved in the study to check the effect on crystal size with cooling rate in the laboratory. In addition to this, it is used in veterinary medicine and also employed for coating pills, in which the medicine is intended for enteric release salol.

Solubility
Soluble in alcohol, ether, chloroform, turpentine, acetone and benzene. Sparingly soluble in chloroformbenzene. Insoluble in water.

Notes
Light sensitive. Incompatible with strong oxidants, strong acids and strong bases. Store in a cool place.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text