Sodium 1-pentanesulfonate, 99% (dry wt.), water <1.5%, Thermo Scientific Chemicals
Sodium 1-pentanesulfonate, 99% (dry wt.), water &lt;1.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium 1-pentanesulfonate, 99% (dry wt.), water <1.5%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number B20699.14
also known as B20699-14
Price (USD)/ Each
130.65
Online exclusive
145.00 
Save 14.35 (10%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
130.65
Online exclusive
145.00 
Save 14.35 (10%)
Add to cart
Sodium 1-pentanesulfonate, 99% (dry wt.), water <1.5%, Thermo Scientific Chemicals
Catalog numberB20699.14
Price (USD)/ Each
130.65
Online exclusive
145.00 
Save 14.35 (10%)
-
Add to cart
Chemical Identifiers
CAS22767-49-3
IUPAC Namesodium pentane-1-sulfonate
Molecular FormulaC5H11NaO3S
InChI KeyROBLTDOHDSGGDT-UHFFFAOYSA-M
SMILES[Na+].CCCCCS([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay (unspecified)≥98.5% (dry wt. basis)
Water Content (Karl Fischer Titration)≤1.5%
Identification (FTIR)Conforms
FormPowder
Sodium 1-pentanesulfonate is used as a base in the Suzuki coupling reaction. It is used as a surfactant in studies on drug release from the water-in -oil-in-water multiple emulsion process in vitro. It is utilized in the analysis of pharmaceutical products, metabolites, small organic molecule compounds, peptides by RP-HPLC and micellar electrokinetic chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium 1-pentanesulfonate is used as a base in the Suzuki coupling reaction. It is used as a surfactant in studies on drug release from the water-in -oil-in-water multiple emulsion process in vitro. It is utilized in the analysis of pharmaceutical products, metabolites, small organic molecule compounds, peptides by RP-HPLC and micellar electrokinetic chromatography.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only

General References:

  1. Peng, G.; Zhu, X.; Chen, J.; Jin, X.; Chen, S.; Wei, D. Rapid Speciation of Lead in Water by High-Performance Liquid Chromatography-Inductively Coupled Plasma Mass Spectrometry. Anal. Lett. 2015, 48 (16), 2639-2649.
  2. Cipriani, S.; Bakshi, R.; Schwarzschild, M. A. Protection by inosine in a cellular model of Parkinson’s disease. Neuroscience 2014, 274, 242-249.