Sodium 1-pentanesulfonate is used as a base in the Suzuki coupling reaction. It is used as a surfactant in studies on drug release from the water-in -oil-in-water multiple emulsion process in vitro. It is utilized in the analysis of pharmaceutical products, metabolites, small organic molecule compounds, peptides by RP-HPLC and micellar electrokinetic chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium 1-pentanesulfonate is used as a base in the Suzuki coupling reaction. It is used as a surfactant in studies on drug release from the water-in -oil-in-water multiple emulsion process in vitro. It is utilized in the analysis of pharmaceutical products, metabolites, small organic molecule compounds, peptides by RP-HPLC and micellar electrokinetic chromatography.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
RUO – Research Use Only
General References:
- Peng, G.; Zhu, X.; Chen, J.; Jin, X.; Chen, S.; Wei, D. Rapid Speciation of Lead in Water by High-Performance Liquid Chromatography-Inductively Coupled Plasma Mass Spectrometry. Anal. Lett. 2015, 48 (16), 2639-2649.
- Cipriani, S.; Bakshi, R.; Schwarzschild, M. A. Protection by inosine in a cellular model of Parkinson’s disease. Neuroscience 2014, 274, 242-249.