Thermo Scientific Chemicals

4-Allyl-1,6-heptadien-4-ol, 99%, Thermo Scientific Chemicals

Catalog number: B20703.09
10 g, Each
Thermo Scientific Chemicals

4-Allyl-1,6-heptadien-4-ol, 99%, Thermo Scientific Chemicals

Catalog number: B20703.09
10 g, Each
Quantity
Catalog number: B20703.09
also known as B20703-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10202-75-2
IUPAC Name
4-(prop-2-en-1-yl)hepta-1,6-dien-4-ol
Molecular Formula
C10H16O
InChI Key
SUXQWOWVXDXQSE-UHFFFAOYSA-N
SMILES
OC(CC=C)(CC=C)CC=C
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥98.5%
Refractive Index
1.4695-1.4725 @ 20?C

Description

A highly diastereoselective synthesis of homoallylic alcohols bearing up to two adjacent quaternary centers was achieved using these polysubstituted allylic reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A highly diastereoselective synthesis of homoallylic alcohols bearing up to two adjacent quaternary centers was achieved using these polysubstituted allylic reagents.

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text