Thermo Scientific Chemicals

Ethylmalonic acid, 97+%, Thermo Scientific Chemicals

Catalog number: B20715.14
25 g, Each
Thermo Scientific Chemicals

Ethylmalonic acid, 97+%, Thermo Scientific Chemicals

Catalog number: B20715.14
25 g, Each
Quantity
Catalog number: B20715.14
also known as B20715-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
601-75-2
IUPAC Name
2-ethylpropanedioic acid
Molecular Formula
C5H8O4
InChI Key
UKFXDFUAPNAMPJ-UHFFFAOYSA-N
SMILES
CCC(C(O)=O)C(O)=O
Assay (Silylated GC)
≥97.0%
Form
Crystals or powder or crystalline powder
Identification (FTIR)
Conforms
Appearance (Color)
White

Description

Ethylmalonic acid is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethylmalonic acid is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in water, methanol (very faint turbidity), and ethanol.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text