Thermo Scientific Chemicals

Methyl 3-mercaptopropionate, 98%, Thermo Scientific Chemicals

Catalog number: B20718.22
100 g, Each
Thermo Scientific Chemicals

Methyl 3-mercaptopropionate, 98%, Thermo Scientific Chemicals

Catalog number: B20718.22
100 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: B20718.22
also known as B20718-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
2935-90-2
IUPAC Name
methyl 3-sulfanylpropanoate
Molecular Formula
C4H8O2S
InChI Key
LDTLDBDUBGAEDT-UHFFFAOYSA-N
SMILES
COC(=O)CCS
Appearance (Color)
Clear colorless
Form
Liquid
Assay (GC)
≥97.5%
Refractive Index
1.4615-1.4655 @ 20?C

Description

Methyl 3-mercaptopropionate is a click chemistry reagent used in the modification of oxazolines using thiol-ene click modification. It also has implications for self-assembled monolayers (SAMs). It is used in the colorimetric detection of mercury(II) in a high-salinity solution using gold nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methyl 3-mercaptopropionate is a click chemistry reagent used in the modification of oxazolines using thiol-ene click modification. It also has implications for self-assembled monolayers (SAMs). It is used in the colorimetric detection of mercury(II) in a high-salinity solution using gold nanoparticles.

Solubility
Soluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text