Thermo Scientific Chemicals

2-Methyl-1-phenyl-1-propanol, 98%, Thermo Scientific Chemicals

Catalog number: B20729.06
5 g, Each
Thermo Scientific Chemicals

2-Methyl-1-phenyl-1-propanol, 98%, Thermo Scientific Chemicals

Catalog number: B20729.06
5 g, Each
Quantity
Catalog number: B20729.06
also known as B20729-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
611-69-8
IUPAC Name
2-methyl-1-phenylpropan-1-ol
Molecular Formula
C10H14O
InChI Key
GMDYDZMQHRTHJA-UHFFFAOYNA-N
SMILES
CC(C)C(O)C1=CC=CC=C1
Assay (GC)
≥97.5%
Appearance (Color)
Clear colorless
Refractive Index
1.5115-1.5155 @ 20?C
Form
Liquid

Description

It is used in the large-scale preparation of (R)-and (S)-1-(2, 4, 6-triisopropylphenyl) ethanol and also in the baker's yeast mediated asymmetric reduction of cinnamaldehyde derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the large-scale preparation of (R)-and (S)-1-(2, 4, 6-triisopropylphenyl) ethanol and also in the baker′s yeast mediated asymmetric reduction of cinnamaldehyde derivatives.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text