Thermo Scientific Chemicals

2-Ethylcyclohexanone, 99%, Thermo Scientific Chemicals

Catalog number: B20748.06
5 g, Each
Thermo Scientific Chemicals

2-Ethylcyclohexanone, 99%, Thermo Scientific Chemicals

Catalog number: B20748.06
5 g, Each
Quantity
Catalog number: B20748.06
also known as B20748-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4423-94-3
IUPAC Name
2-ethylcyclohexan-1-one
Molecular Formula
C8H14O
InChI Key
WKYYYUWKFPFVEY-UHFFFAOYNA-N
SMILES
CCC1CCCCC1=O
Appearance (Color)
Clear colorless to pale yellow
Form
Liquid
Assay (GC)
≥98.5%
Refractive Index
1.4500-1.4530 @ 20°C

Description

2-Ethylcyclohexanone is used as a pharmaceutical intermediate. 2-Methyl and 2-ethyl derivatives of cyclohexanone were allowed to react with deuterium in tBuOD using platinum group metals as catalysts to study the substituent effects in heterogeneous catalysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Ethylcyclohexanone is used as a pharmaceutical intermediate. 2-Methyl and 2-ethyl derivatives of cyclohexanone were allowed to react with deuterium in tBuOD using platinum group metals as catalysts to study the substituent effects in heterogeneous catalysis.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text