Thermo Scientific Chemicals

Sodium deoxycholate monohydrate, 98%, Thermo Scientific Chemicals

Catalog number: B20759.36
500 g, Each
Thermo Scientific Chemicals

Sodium deoxycholate monohydrate, 98%, Thermo Scientific Chemicals

Catalog number: B20759.36
500 g, Each
Quantity
Catalog number: B20759.36
also known as B20759-36
Price (USD)
Have Questions?

Chemical Identifiers

CAS
145224-92-6
IUPAC Name
sodium (4R)-4-[(1R,3aS,3bR,5aR,7R,9aS,9bS,11S,11aR)-7,11-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanoate hydrate
Molecular Formula
C24H41NaO5
InChI Key
NDVHNZISGVSFMP-WTCAICSISA-M
SMILES
O.[Na+].[H][C@@]1(CC[C@@]2([H])[C@]3([H])CC[C@]4([H])C[C@H](O)CC[C@]4(C)[C@@]3([H])C[C@H](O)[C@]12C)[C@H](C)CCC([O-])=O
Appearance (Color)
White
Optical Rotation
44.0 ± 2.0° (C=2 in water, UK-sourced material)
Form
Powder
Assay from Supplier's CofA
≥97.5% (HPLC, US-sourced material)
Identification (FTIR)
Conforms (UK-sourced material)

Description

Sodium deoxycholate monohydrate is used as a solubilization agent for estrogen synthetase from human placental microsomes. It is also used to study the immobilization of lipoxygenase in an alginate-silicate gel matrix and to investigate the in vitro dissolution of gall stones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium deoxycholate monohydrate is used as a solubilization agent for estrogen synthetase from human placental microsomes. It is also used to study the immobilization of lipoxygenase in an alginate-silicate gel matrix and to investigate the in vitro dissolution of gall stones.

Solubility
Soluble in water.

Notes
Incompatible with oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text