Benzalkonium chloride is a cationic surface active agent.It is used as a preservative in pharmaceutical products like eye, ear and nasal drops. It finds application in personal care products such as wet wipes, shampoos, hand sanitizers, deodorants and cosmetics. It is widely used in the chemical industry as a precipitant and emulsifier/de-emulsifier. It acts as corrosion inhibitor in the oil and gas industry pipeline by preventing the formation of sulfurous gases. It is also used in various non-consumer process like surgical disinfection.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Benzalkonium chloride is a cationic surface active agent.It is used as a preservative in pharmaceutical products like eye, ear and nasal drops. It finds application in personal care products such as wet wipes, shampoos, hand sanitizers, deodorants and cosmetics. It is widely used in the chemical industry as a precipitant and emulsifier/de-emulsifier. It acts as corrosion inhibitor in the oil and gas industry pipeline by preventing the formation of sulfurous gases. It is also used in various non-consumer process like surgical disinfection.
Solubility
Miscible with water, methanol and acetone.
Notes
Incompatible with strong oxidizing agents, strong acids, strong bases and metals.
RUO – Research Use Only
General References:
- Sabatini, C.; Pashley, D. H. Aging of adhesive interfaces treated with benzalkonium chloride and benzalkonium methacrylate. Eur J Oral Sci. 2015, 123 (2), 102-107.
- Kim, J. M.; Ahn, Y.; LiPuma, J. J.; Hussong, D.; Cerniglia, C. E. Survival and susceptibility of Burkholderia cepacia complex in chlorhexidine gluconate and benzalkonium chloride. J. Ind. Microbiol. Biotechnol. 2015, 42 (6), 905-913.