Dimethyl 5-aminoisophthalate, 98%, Thermo Scientific Chemicals
Dimethyl 5-aminoisophthalate, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Dimethyl 5-aminoisophthalate, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number B20761.14
also known as B20761-14
Price (USD)/ Each
41.10
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
41.10
Add to cart
Dimethyl 5-aminoisophthalate, 98%, Thermo Scientific Chemicals
Catalog numberB20761.14
Price (USD)/ Each
41.10
-
Add to cart
Chemical Identifiers
CAS99-27-4
IUPAC Name1,3-dimethyl 5-aminobenzene-1,3-dicarboxylate
Molecular FormulaC10H11NO4
InChI KeyDEKPYXUDJRABNK-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC(=CC(N)=C1)C(=O)OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream or pale yellow to pale brown
FormCrystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)≥97.5 to ≤102.5%
Assay (GC)≥97.5%
Identification (FTIR)Conforms
View more
Dimethyl 5-aminoisophthalate is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. Hexagonal assembly of double helices of luminescent calcium 5-aminoisophthalate coordination polymer leaves enough room for the formation of loosely held water pipes, which could be dehydrated at very low temperatures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl 5-aminoisophthalate is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. Hexagonal assembly of double helices of luminescent calcium 5-aminoisophthalate coordination polymer leaves enough room for the formation of loosely held water pipes, which could be dehydrated at very low temperatures.

Solubility
Soluble in Chloroform.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only
Ramaswamy Murugavel .;Parimal Kumar.; Mrinalini G. Walawalkar.; Ramyaa Mathialagan. A Double Helix Is the Repeating Unit in a Luminescent Calcium 5-Aminoisophthalate Supramolecular Edifice with Water-Filled Hexagonal Channels.Inorg. Chem. 2007, 46 (17),6828-6830 .