Thermo Scientific Chemicals

Dimethyl 5-aminoisophthalate, 98%, Thermo Scientific Chemicals

Catalog number: B20761.22
100 g, Each
Thermo Scientific Chemicals

Dimethyl 5-aminoisophthalate, 98%, Thermo Scientific Chemicals

Catalog number: B20761.22
100 g, Each
Quantity
Catalog number: B20761.22
also known as B20761-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
99-27-4
IUPAC Name
1,3-dimethyl 5-aminobenzene-1,3-dicarboxylate
Molecular Formula
C10H11NO4
InChI Key
DEKPYXUDJRABNK-UHFFFAOYSA-N
SMILES
COC(=O)C1=CC(=CC(N)=C1)C(=O)OC
Appearance (Color)
White to cream or pale yellow to pale brown
Form
Crystals or powder or crystalline powder
Assay (Non-aqueous acid-base Titration)
≥97.5 to ≤102.5%
Assay (GC)
≥97.5%
Identification (FTIR)
Conforms

Description

Dimethyl 5-aminoisophthalate is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. Hexagonal assembly of double helices of luminescent calcium 5-aminoisophthalate coordination polymer leaves enough room for the formation of loosely held water pipes, which could be dehydrated at very low temperatures.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethyl 5-aminoisophthalate is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. Hexagonal assembly of double helices of luminescent calcium 5-aminoisophthalate coordination polymer leaves enough room for the formation of loosely held water pipes, which could be dehydrated at very low temperatures.

Solubility
Soluble in Chloroform.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text